Product Details

SNP ID
rs62057932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66935460 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCACCTATGACTGCTCAGTCCC[A/G]CTCTCCTACCACACCCACCTTTCCC
Phenotype
MIM: 605278 MIM: 614778 MIM: 179503
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CES2 PubMed Links

Gene Details

Gene
CES2
Gene Name
carboxylesterase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003869.5 1017 Missense Mutation CAC,CGC H6R NP_003860.2
NM_198061.2 1017 Missense Mutation CAC,CGC H6R NP_932327.1
XM_011523421.2 1017 UTR 5 XP_011521723.1
XM_017023818.1 1017 UTR 5 XP_016879307.1
Gene
FAM96B
Gene Name
family with sequence similarity 96 member B
There are no transcripts associated with this gene.

Gene
RRAD
Gene Name
RRAD, Ras related glycolysis inhibitor and calcium channel regulator
There are no transcripts associated with this gene.

View Full Product Details