Product Details

SNP ID
rs62059744
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50642447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTCTCACAGCCCCAAAGTCCCC[C/T]GGTTGTTGCCACAGAGCAGGTGTCT
Phenotype
MIM: 604323
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCC3 PubMed Links

Gene Details

Gene
ABCC3
Gene Name
ATP binding cassette subfamily C member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144070.1 Intron NP_001137542.1
NM_003786.3 Intron NP_003777.2
XM_005257763.3 Intron XP_005257820.1
XM_011525422.2 Intron XP_011523724.1
XM_011525423.1 Intron XP_011523725.1
XM_011525424.2 Intron XP_011523726.1
XM_011525425.2 Intron XP_011523727.1
XM_017025265.1 Intron XP_016880754.1
XM_017025266.1 Intron XP_016880755.1

View Full Product Details