Product Details

SNP ID
rs61756762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:40861389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTGATCACTTTCAATAATTTT[A/C]TTTGTATTAATATTTCTGCCATTAA
Phenotype
MIM: 611336 MIM: 606796 MIM: 613553
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DNAJB7 PubMed Links

Gene Details

Gene
DNAJB7
Gene Name
DnaJ heat shock protein family (Hsp40) member B7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145174.1 738 Missense Mutation AAG,AAT K202N NP_660157.1
Gene
ST13
Gene Name
suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein)
There are no transcripts associated with this gene.

Gene
XPNPEP3
Gene Name
X-prolyl aminopeptidase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204827.1 738 Intron NP_001191756.1
NM_022098.3 738 Intron NP_071381.1

View Full Product Details