Product Details

SNP ID
rs61280166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:88260904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTACTTTATATACAAATTGCCATA[C/T]AGCAATTTTTAACCTATTTCCTCAT
Phenotype
MIM: 611065
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPM1K PubMed Links

Gene Details

Gene
PPM1K
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152542.4 2534 UTR 3 NP_689755.3
XM_005262775.3 2534 Intron XP_005262832.1
XM_006714111.3 2534 UTR 3 XP_006714174.1
XM_017007803.1 2534 UTR 3 XP_016863292.1
XM_017007804.1 2534 UTR 3 XP_016863293.1
XM_017007805.1 2534 UTR 3 XP_016863294.1
XM_017007806.1 2534 UTR 3 XP_016863295.1
XM_017007807.1 2534 UTR 3 XP_016863296.1

View Full Product Details