Product Details

SNP ID
rs59644448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:104065667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATATGAAGGTAAAAAGTTACTGAC[A/G]CTTTGGTCTTAGTGTAAAATCCCTG
Phenotype
MIM: 607926
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLT8D2 PubMed Links

Gene Details

Gene
GLT8D2
Gene Name
glycosyltransferase 8 domain containing 2
There are no transcripts associated with this gene.

Gene
HCFC2
Gene Name
host cell factor C2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013320.2 Intron NP_037452.1
XM_017019241.1 Intron XP_016874730.1
XM_017019242.1 Intron XP_016874731.1

View Full Product Details