Product Details

SNP ID
rs56268528
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:206904986 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGAGACTCCTTGGCACCACAGTC[C/T]GAGCCTGGGGTTGGGGGATAGCTGG
Phenotype
MIM: 606015 MIM: 604136
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FCMR PubMed Links

Gene Details

Gene
FCMR
Gene Name
Fc fragment of IgM receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142473.1 1326 UTR 3 NP_001135945.1
NM_001193338.1 1326 UTR 3 NP_001180267.1
NM_005449.4 1326 UTR 3 NP_005440.1
XM_005273351.3 1326 UTR 3 XP_005273408.1
XM_005273352.4 1326 UTR 3 XP_005273409.1
Gene
IL24
Gene Name
interleukin 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185156.1 1326 Intron NP_001172085.1
NM_001185157.1 1326 Intron NP_001172086.1
NM_001185158.1 1326 Intron NP_001172087.1
NM_006850.3 1326 Intron NP_006841.1
XM_011509101.2 1326 Intron XP_011507403.1
XM_017000121.1 1326 Intron XP_016855610.1

View Full Product Details