Product Details

SNP ID
rs57043559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:63883228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAGTAGAGGCTCAGTAAATGTTT[A/C]TTGAATATAAATCTGAATCAGACCA
Phenotype
MIM: 607640
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATXN7 PubMed Links

Gene Details

Gene
ATXN7
Gene Name
ataxin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000333.3 Intron NP_000324.1
NM_001128149.2 Intron NP_001121621.2
NM_001177387.1 Intron NP_001170858.1

View Full Product Details