Product Details
- SNP ID
-
rs57936737
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:168103536 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATACTTCAATATTGTTACCTAATA[G/T]CAGAGTTTCATTAGTACTACGACTT
- Phenotype
-
MIM: 608008
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ANXA10
PubMed Links
Gene Details
- Gene
- ANXA10
- Gene Name
- annexin A10
View Full Product Details