Product Details

SNP ID
rs56213863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:111299142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGACCACATAATTCCTTTTTTA[C/T]TTTACACAGTTATACAAATATTCTA
Phenotype
MIM: 602776
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
REV3L PubMed Links

Gene Details

Gene
REV3L
Gene Name
REV3 like, DNA directed polymerase zeta catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286431.1 10897 UTR 3 NP_001273360.1
NM_001286432.1 10897 UTR 3 NP_001273361.1
NM_002912.4 10897 UTR 3 NP_002903.3
XM_011536028.1 10897 UTR 3 XP_011534330.1
XM_011536029.2 10897 Intron XP_011534331.1
XM_011536030.2 10897 UTR 3 XP_011534332.1
XM_011536031.2 10897 Intron XP_011534333.1
XM_011536032.2 10897 Intron XP_011534334.1
XM_011536036.2 10897 Intron XP_011534338.1
XM_017011152.1 10897 Intron XP_016866641.1
XM_017011153.1 10897 Intron XP_016866642.1
XM_017011154.1 10897 Intron XP_016866643.1
XM_017011155.1 10897 Intron XP_016866644.1

View Full Product Details