Product Details

SNP ID
rs61759411
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74941960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTGCCACTGCCTTCAGCTGGGA[C/T]GCACAGGCTCCTCCCCTGGTGACCT
Phenotype
MIM: 601121
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PGF PubMed Links

Gene Details

Gene
PGF
Gene Name
placental growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207012.1 1781 UTR 3 NP_001193941.1
NM_001293643.1 1781 UTR 3 NP_001280572.1
NM_002632.5 1781 UTR 3 NP_002623.2

View Full Product Details