Product Details

SNP ID
rs61914248
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52276845 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCGCTGGCTCAAGTCTCACTCCA[C/T]GGAGGGAAGGGCTTCTGTTCCCGGT
Phenotype
MIM: 602153 MIM: 601928
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT81 PubMed Links

Gene Details

Gene
KRT81
Gene Name
keratin 81
There are no transcripts associated with this gene.

Gene
KRT86
Gene Name
keratin 86
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320198.1 Intron NP_001307127.1
XM_005268866.4 Intron XP_005268923.1
XM_017019296.1 Intron XP_016874785.1

View Full Product Details