Product Details

SNP ID
rs56289122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:41681112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGGACAGCTCTGTAGTACTCAA[C/T]TTCCTCCTGCCTCGATTTCCTCATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00111 PubMed Links

Gene Details

Gene
LINC00111
Gene Name
long intergenic non-protein coding RNA 111
There are no transcripts associated with this gene.

View Full Product Details