Product Details
- SNP ID
-
rs60633545
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:19943842 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGAAAATTCAGGGTAAAATTCAG[A/G]TGATTCATACAGGATTTATTTTTCC
- Phenotype
-
MIM: 609708
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LPL
PubMed Links
Gene Details
- Gene
- LPL
- Gene Name
- lipoprotein lipase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000237.2 |
|
Intron |
|
|
NP_000228.1 |
View Full Product Details