Product Details
- SNP ID
-
rs55665437
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
14
- Location
-
Chr.1:93993101 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTTTCTGAATTCGCTGCATGCTC[C/T]TCGTGTGTTTGTTTTCTGCTGCAGT
- Phenotype
-
MIM: 601691
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ABCA4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76201551] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABCA4
- Gene Name
- ATP binding cassette subfamily A member 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000350.2 |
7062 |
UTR 3 |
|
|
NP_000341.2 |
View Full Product Details