Product Details

SNP ID
rs55921233
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:97077882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCATAGAAATAAGCCATAGATTG[C/T]TACTTTCTGATTTACTACTTATAGA
Phenotype
MIM: 612779
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPYD PubMed Links

Gene Details

Gene
DPYD
Gene Name
dihydropyrimidine dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000110.3 4519 UTR 3 NP_000101.2
NM_001160301.1 4519 Intron NP_001153773.1
XM_005270562.3 4519 UTR 3 XP_005270619.2
XM_006710397.3 4519 Intron XP_006710460.1
XM_017000507.1 4519 UTR 3 XP_016855996.1
XM_017000508.1 4519 UTR 3 XP_016855997.1
XM_017000509.1 4519 UTR 3 XP_016855998.1
XM_017000510.1 4519 UTR 3 XP_016855999.1

View Full Product Details