Product Details

SNP ID
rs61689775
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:77217108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCATGCTTGCCAGGCTTCAAAGG[G/T]GTGACAGCATTCTTGATAGGTAGTA
Phenotype
MIM: 276903
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GDPD4 PubMed Links

Gene Details

Gene
GDPD4
Gene Name
glycerophosphodiester phosphodiesterase domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182833.1 2178 UTR 3 NP_878253.1
XM_011544826.1 2178 UTR 3 XP_011543128.1
XM_011544833.1 2178 Intron XP_011543135.1
XM_011544834.1 2178 Silent Mutation ACA,ACC T593T XP_011543136.1
XM_017017348.1 2178 Intron XP_016872837.1
XM_017017349.1 2178 Intron XP_016872838.1
XM_017017350.1 2178 Intron XP_016872839.1
XM_017017351.1 2178 Intron XP_016872840.1
Gene
MYO7A
Gene Name
myosin VIIA
There are no transcripts associated with this gene.

View Full Product Details