Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001042410.1 | 64 | UTR 5 | NP_001035869.1 | ||
NM_001282792.1 | 64 | Intron | NP_001269721.1 | ||
NM_018089.2 | 64 | Intron | NP_060559.2 | ||
XM_005246663.2 | 64 | Intron | XP_005246720.1 | ||
XM_011511392.2 | 64 | Intron | XP_011509694.1 | ||
XM_017004420.1 | 64 | Intron | XP_016859909.1 | ||
XM_017004421.1 | 64 | Intron | XP_016859910.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001077198.2 | 64 | Intron | NP_001070666.1 | ||
NM_024085.4 | 64 | Intron | NP_076990.4 |