Product Details

SNP ID
rs73886010
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:186667033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAACTGGCCACTTGCATACTTA[A/G]AATTCACAAAGGCTGTGCAGAATGC
Phenotype
MIM: 142640
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HRG PubMed Links

Gene Details

Gene
HRG
Gene Name
histidine rich glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000412.4 Intron NP_000403.1
XM_005247415.3 Intron XP_005247472.1

View Full Product Details