Product Details

SNP ID
rs73444905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:55437523 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAAGAGGATGGAGGATATATGG[A/G]GCACTGTATGCAGGCAGAGAGAAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HMGCLL1 PubMed Links

Gene Details

Gene
HMGCLL1
Gene Name
3-hydroxymethyl-3-methylglutaryl-CoA lyase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042406.1 Intron NP_001035865.1
NM_001287741.1 Intron NP_001274670.1
NM_001287746.1 Intron NP_001274675.1
NM_001287753.1 Intron NP_001274682.1
NM_019036.2 Intron NP_061909.2
XM_011514700.2 Intron XP_011513002.1

View Full Product Details