Product Details

SNP ID
rs73645468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35562896 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTTGTTCCACCACCTCCAGTGT[C/T]GGCTCCTCCAGCTGCCCCTCCGGCT
Phenotype
MIM: 611053
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM166B PubMed Links

Gene Details

Gene
FAM166B
Gene Name
family with sequence similarity 166 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099951.3 542 Silent Mutation CCA,CCG P157P NP_001093421.1
NM_001164310.2 542 Silent Mutation CCA,CCG P157P NP_001157782.1
NM_001287238.1 542 Silent Mutation CCA,CCG P157P NP_001274167.1
NM_001287239.1 542 Silent Mutation CCA,CCG P157P NP_001274168.1
XM_011518028.2 542 Silent Mutation CCA,CCG P152P XP_011516330.1
Gene
RUSC2
Gene Name
RUN and SH3 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135999.1 542 Intron NP_001129471.1
XM_006716896.2 542 Intron XP_006716959.1

View Full Product Details