Product Details

SNP ID
rs73442560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:72402317 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGTCCAATTCAATGACTTCAAAC[G/T]TTTACCTACTGGATTGCAAGAATTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC105370888 PubMed Links

Gene Details

Gene
LOC105370888
Gene Name
uncharacterized LOC105370888
There are no transcripts associated with this gene.

Gene
TMEM202
Gene Name
transmembrane protein 202
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080462.1 Intron NP_001073931.1
XM_011521497.1 Intron XP_011519799.1
XM_011521499.1 Intron XP_011519801.1

View Full Product Details