Product Details

SNP ID
rs68091935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:81991022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAGGCAAAGAGCCATCAGCAGC[A/G]AATAATACTTGGGCTCTGCTCTCAA
Phenotype
MIM: 616164
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SDR42E1 PubMed Links

Gene Details

Gene
SDR42E1
Gene Name
short chain dehydrogenase/reductase family 42E, member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145168.2 9402 Intron NP_660151.2
XM_005256257.3 9402 UTR 3 XP_005256314.1
XM_011523471.2 9402 UTR 3 XP_011521773.1
XM_011523472.2 9402 UTR 3 XP_011521774.1

View Full Product Details