Product Details

SNP ID
rs72898209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78160963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTTTCATCTCCCAGAAATTACC[C/T]CTGTGGTCTCCATTGCCTACAAAGT
Phenotype
MIM: 603926
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C17orf99 PubMed Links

Gene Details

Gene
C17orf99
Gene Name
chromosome 17 open reading frame 99
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163075.1 145 Missense Mutation CCT,TCT P27S NP_001156547.1
XM_011524152.2 145 Missense Mutation CCT,TCT P31S XP_011522454.1
XM_017023997.1 145 Missense Mutation CCT,TCT P31S XP_016879486.1
XM_017023998.1 145 Missense Mutation CCT,TCT P27S XP_016879487.1
XM_017023999.1 145 Intron XP_016879488.1
XM_017024000.1 145 UTR 5 XP_016879489.1
XM_017024001.1 145 UTR 5 XP_016879490.1
Gene
SYNGR2
Gene Name
synaptogyrin 2
There are no transcripts associated with this gene.

View Full Product Details