Product Details

SNP ID
rs73366211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82235154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGGGCTGGCATGGTTTGCTCTC[A/G]TGGGCTGGATGGGAGCCTCCCTAGC
Phenotype
MIM: 600864 MIM: 603877
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSNK1D PubMed Links

Gene Details

Gene
CSNK1D
Gene Name
casein kinase 1 delta
There are no transcripts associated with this gene.

Gene
MIR6787
Gene Name
microRNA 6787
There are no transcripts associated with this gene.

Gene
SLC16A3
Gene Name
solute carrier family 16 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042422.2 1468 Intron NP_001035887.1
NM_001042423.2 1468 Intron NP_001035888.1
NM_001206950.1 1468 Intron NP_001193879.1
NM_001206951.1 1468 Intron NP_001193880.1
NM_001206952.1 1468 Intron NP_001193881.1
NM_004207.3 1468 Intron NP_004198.1
XM_011523607.1 1468 UTR 5 XP_011521909.1

View Full Product Details