Product Details

SNP ID
rs73537681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38868893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCAGTGAGGCTTTCTCTGATAC[C/G]CCTGTCTAAAACTGCAAAGCCTCCT
Phenotype
MIM: 604480
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RINL PubMed Links

Gene Details

Gene
RINL
Gene Name
Ras and Rab interactor like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195833.1 1977 UTR 3 NP_001182762.1
NM_198445.3 1977 Intron NP_940847.1
XM_006723015.2 1977 UTR 3 XP_006723078.1
XM_006723016.2 1977 UTR 3 XP_006723079.1
XM_011526454.2 1977 UTR 3 XP_011524756.1
XM_011526455.2 1977 UTR 3 XP_011524757.1
Gene
SIRT2
Gene Name
sirtuin 2
There are no transcripts associated with this gene.

View Full Product Details