Product Details

SNP ID
rs73915589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62126141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGAAATGCTGTGGAAAAGGCAG[C/T]TCTCACCATACTGGAAACTTGCACC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LSM14B PubMed Links

Gene Details

Gene
LSM14B
Gene Name
LSM family member 14B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144703.2 Intron NP_653304.2
XM_005260302.3 Intron XP_005260359.1
XM_011528605.2 Intron XP_011526907.1
XM_011528606.2 Intron XP_011526908.1
XM_011528607.2 Intron XP_011526909.1
XM_011528608.2 Intron XP_011526910.1
XM_011528609.2 Intron XP_011526911.1
XM_011528611.2 Intron XP_011526913.1
XM_011528612.2 Intron XP_011526914.1
XM_011528613.2 Intron XP_011526915.1
XM_011528614.2 Intron XP_011526916.1
XM_011528615.2 Intron XP_011526917.1
XM_017027688.1 Intron XP_016883177.1
XM_017027689.1 Intron XP_016883178.1
XM_017027690.1 Intron XP_016883179.1

View Full Product Details