Product Details

SNP ID
rs72660611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:118040387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCTCATCTAAGGAAAGGGGGAGG[A/G]GTGAAAAGAAGCCCAAGGACATGGT
Phenotype
MIM: 603950
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NDST3 PubMed Links

Gene Details

Gene
NDST3
Gene Name
N-deacetylase and N-sulfotransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004784.2 Intron NP_004775.1
XM_006714416.3 Intron XP_006714479.1
XM_011532415.2 Intron XP_011530717.1
XM_011532416.2 Intron XP_011530718.1
XM_017008839.1 Intron XP_016864328.1
XM_017008840.1 Intron XP_016864329.1
XM_017008841.1 Intron XP_016864330.1
XM_017008842.1 Intron XP_016864331.1
XM_017008843.1 Intron XP_016864332.1
XM_017008844.1 Intron XP_016864333.1

View Full Product Details