Product Details

SNP ID
rs73858517
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:158667130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCATGTTCAAATTTCCTTCAGA[C/T]AAAAAGAACAGAAAGGTCAGAATCT
Phenotype
MIM: 616210 MIM: 231675
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
C4orf46 PubMed Links

Gene Details

Gene
C4orf46
Gene Name
chromosome 4 open reading frame 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008393.3 3074 UTR 3 NP_001008394.1
Gene
ETFDH
Gene Name
electron transfer flavoprotein dehydrogenase
There are no transcripts associated with this gene.

View Full Product Details