Product Details
- SNP ID
-
rs67694373
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:165384314 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTTTCAAATCTCCAAGTTAGATTC[A/G]TACTGTATAAGATTTCCCAGGCTTT
- Phenotype
-
MIM: 114855
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CPE
PubMed Links
Gene Details
- Gene
- CPE
- Gene Name
- carboxypeptidase E
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001873.3 |
|
Intron |
|
|
NP_001864.1 |
- Gene
- MIR578
- Gene Name
- microRNA 578
There are no transcripts associated with this gene.
View Full Product Details