Product Details

SNP ID
rs72639212
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:11847213 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCTCCGAGGGGCAGTGAGCAGC[G/A]CCCTCAGCTTGCTTTTTAGGAGGGC
Phenotype
MIM: 602726 MIM: 108780
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CLCN6 PubMed Links

Gene Details

Gene
CLCN6
Gene Name
chloride voltage-gated channel 6
There are no transcripts associated with this gene.

Gene
NPPA
Gene Name
natriuretic peptide A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006172.3 449 Missense Mutation NP_006163.1
Gene
NPPA-AS1
Gene Name
NPPA antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details