Product Details

SNP ID
rs72943019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:105252239 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGACTTTAGATGCTTTATTACA[C/G]CATAACAAGAGAGTCACTAAGCTCA
Phenotype
MIM: 604838 MIM: 606237
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GPR45 PubMed Links

Gene Details

Gene
GPR45
Gene Name
G protein-coupled receptor 45
There are no transcripts associated with this gene.

Gene
TGFBRAP1
Gene Name
transforming growth factor beta receptor associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142621.2 Intron NP_001136093.1
NM_004257.5 Intron NP_004248.2
XM_011512193.2 Intron XP_011510495.1
XM_017005339.1 Intron XP_016860828.1

View Full Product Details