Product Details

SNP ID
rs72724960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:188091588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAGGAAAACGCCAACTGTGTCCA[A/G]CTTCTTTTCCAGGAAGAGCCTTTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIML2 PubMed Links

Gene Details

Gene
TRIML2
Gene Name
tripartite motif family like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303419.1 1393 Silent Mutation CTG,TTG L392L NP_001290348.1
NM_173553.2 1393 UTR 3 NP_775824.2
XM_011531737.2 1393 Silent Mutation CTG,TTG L367L XP_011530039.1
XM_011531738.1 1393 Silent Mutation CTG,TTG L367L XP_011530040.1
XM_011531739.1 1393 Silent Mutation CTG,TTG L275L XP_011530041.1
XM_017007882.1 1393 Silent Mutation CTG,TTG L367L XP_016863371.1

View Full Product Details