Product Details

SNP ID
rs73520932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109464669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTAGATGAAGATTAGCTGGGTGT[G/T]GTGGTACATCCCTGTAGTCCCAGCT
Phenotype
MIM: 607129 MIM: 614064
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MICAL1 PubMed Links

Gene Details

Gene
MICAL1
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159291.1 3444 Intron NP_001152763.1
NM_001286613.1 3444 Intron NP_001273542.1
NM_022765.3 3444 Intron NP_073602.3
Gene
ZBTB24
Gene Name
zinc finger and BTB domain containing 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164313.1 3444 Intron NP_001157785.1
NM_014797.2 3444 UTR 3 NP_055612.2

View Full Product Details