Product Details

SNP ID
rs73112143
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49297467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGAGGAGGAGCTGCGACAGCTAAA[A/G]GAGGAGATGGCGCGGCACCTGAGGG
Phenotype
MIM: 170710
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LOC101927267 PubMed Links

Gene Details

Gene
LOC101927267
Gene Name
uncharacterized LOC101927267
There are no transcripts associated with this gene.

Gene
PRPH
Gene Name
peripherin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006262.3 1295 Silent Mutation AAA,AAG K369K NP_006253.2
XM_005269025.1 1295 Silent Mutation AAA,AAG K369K XP_005269082.1

View Full Product Details