Product Details

SNP ID
rs72466468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:113904931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCTCCTTTTTGGGATCTGCCCA[A/G]GAATGTTGTCAAGTGTAATGGCTCC
Phenotype
MIM: 604654
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HTR3B PubMed Links

Gene Details

Gene
HTR3B
Gene Name
5-hydroxytryptamine receptor 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006028.4 136 UTR 5 NP_006019.1
XM_011543063.1 136 Intron XP_011541365.1
XM_017018552.1 136 Intron XP_016874041.1

View Full Product Details