Product Details

SNP ID
rs78314980
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:153969609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGACCAAGGAGACATGAACAGT[G/T]GAACTATTTTTTATTTTTTGAGCCA
Phenotype
MIM: 607138 MIM: 604671 MIM: 604740
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CREB3L4 PubMed Links

Gene Details

Gene
CREB3L4
Gene Name
cAMP responsive element binding protein 3 like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001255978.1 Intron NP_001242907.1
NM_001255979.1 Intron NP_001242908.1
NM_001255980.1 Intron NP_001242909.1
NM_001255981.1 Intron NP_001242910.1
NM_130898.3 Intron NP_570968.1
XM_006711172.2 Intron XP_006711235.1
XM_017000372.1 Intron XP_016855861.1
Gene
JTB
Gene Name
jumping translocation breakpoint
There are no transcripts associated with this gene.

Gene
MIR6737
Gene Name
microRNA 6737
There are no transcripts associated with this gene.

Gene
SLC39A1
Gene Name
solute carrier family 39 member 1
There are no transcripts associated with this gene.

View Full Product Details