Product Details

SNP ID
rs78921047
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:154570350 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCAAACACATCTGGCTCCCAGA[C/T]GTGGTCCTGTACAACAAGTAGGTGC
Phenotype
MIM: 118507
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHRNB2 PubMed Links

Gene Details

Gene
CHRNB2
Gene Name
cholinergic receptor nicotinic beta 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000748.2 594 Silent Mutation GAC,GAT D116D NP_000739.1
XM_017000180.1 594 UTR 5 XP_016855669.1

View Full Product Details