Product Details

SNP ID
rs78346899
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:155235716 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGTGGTAGAACATGGGCTGTTT[G/A]TAAAACGTGTCCTTGGTGATGTCTA
Phenotype
MIM: 606463
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
GBA PubMed Links

Gene Details

Gene
GBA
Gene Name
glucosylceramidase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000157.3 1778 Silent Mutation TAC,TAT Y451Y NP_000148.2
NM_001005741.2 1778 Silent Mutation TAC,TAT Y451Y NP_001005741.1
NM_001005742.2 1778 Silent Mutation TAC,TAT Y451Y NP_001005742.1
NM_001171811.1 1778 Silent Mutation TAC,TAT Y364Y NP_001165282.1
NM_001171812.1 1778 Silent Mutation TAC,TAT Y402Y NP_001165283.1
Gene
GBAP1
Gene Name
glucosylceramidase beta pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details