Product Details

SNP ID
rs173278
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:149346670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCACAACTCCCCAACCCTATTT[C/T]TTGGTCTAAGCTTTGGCCATTTTCC
Phenotype
MIM: 614410
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AFAP1L1 PubMed Links

Gene Details

Gene
AFAP1L1
Gene Name
actin filament associated protein 1 like 1
There are no transcripts associated with this gene.

Gene
GRPEL2
Gene Name
GrpE like 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152407.3 Intron NP_689620.2
Gene
GRPEL2-AS1
Gene Name
GRPEL2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details