Product Details

SNP ID
rs550894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65444469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGAGCCTCTGCTTCCCATCTGGA[A/C]CCTGCTGGGCAGGGCTTCTGAGCTC
Phenotype
MIM: 612769
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC101927789 PubMed Links

Gene Details

Gene
LOC101927789
Gene Name
putative ubiquitin-like protein FUBI-like protein ENSP00000310146
There are no transcripts associated with this gene.

Gene
MIR612
Gene Name
microRNA 612
There are no transcripts associated with this gene.

Gene
NEAT1
Gene Name
nuclear paraspeckle assembly transcript 1 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details