Product Details

SNP ID
rs3890113
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:149476254 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGCCACTGAGAGGCCATAAGAAC[G/T]TGGACAAGTCATTTCCCTTTTCTGG
Phenotype
MIM: 606567
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TM4SF4 PubMed Links

Gene Details

Gene
TM4SF4
Gene Name
transmembrane 4 L six family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004617.3 Intron NP_004608.1

View Full Product Details