Product Details
- SNP ID
-
rs923661
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:132208366 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CGTGAGCATGCTGGCGAGGCTCGGC[A/G]TGTTTCCAGCCAGAAGCCGAAAGGC
- Phenotype
-
MIM: 608407
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DPYSL4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75612245] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DPYSL4
- Gene Name
- dihydropyrimidinase like 4
There are no transcripts associated with this gene.
- Gene
- STK32C
- Gene Name
- serine/threonine kinase 32C
View Full Product Details