Product Details

SNP ID
rs17853024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74952274 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCCGGCGCCCGACTCACTTGTC[A/G]ATGCAGATCTTCTCCACCTGCGGAA
Phenotype
MIM: 605752 MIM: 607828
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HID1 PubMed Links

Gene Details

Gene
HID1
Gene Name
HID1 domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030630.2 2283 Silent Mutation ATC,ATT I713I NP_085133.1
XM_005257226.1 2283 Silent Mutation ATC,ATT I712I XP_005257283.1
Gene
OTOP3
Gene Name
otopetrin 3
There are no transcripts associated with this gene.

View Full Product Details