Product Details

SNP ID
rs910869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34704973 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGGACTCCAGGTGGTTGGGACAG[C/T]ACCAGATGGTGGTGGGATCCCCTCC
Phenotype
MIM: 605299
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NCOA6 PubMed Links

Gene Details

Gene
NCOA6
Gene Name
nuclear receptor coactivator 6
There are no transcripts associated with this gene.

Gene
TP53INP2
Gene Name
tumor protein p53 inducible nuclear protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005260476.2 Intron XP_005260533.1
XM_006723841.2 Intron XP_006723904.1
XM_011528952.2 Intron XP_011527254.1
XM_011528953.2 Intron XP_011527255.1

View Full Product Details