Product Details

SNP ID
rs11621203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74241441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATCTGCCCGGGGGCCTTGGCGTC[C/G]GCACCGGTTGAGCAGGATGAGTGGC
Phenotype
MIM: 142993
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
VSX2 PubMed Links

Gene Details

Gene
VSX2
Gene Name
visual system homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182894.2 Intron NP_878314.1

View Full Product Details