Product Details

SNP ID
rs11818389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133364815 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGGAAGCCCTCCTCACTGACCCA[C/T]GCACAACTGTGCTTTCGACGTGGCC
Phenotype
MIM: 602292
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ECHS1 PubMed Links

Gene Details

Gene
ECHS1
Gene Name
enoyl-CoA hydratase, short chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004092.3 Intron NP_004083.3
Gene
FUOM
Gene Name
fucose mutarotase
There are no transcripts associated with this gene.

Gene
MIR3944
Gene Name
microRNA 3944
There are no transcripts associated with this gene.

View Full Product Details