Product Details
- SNP ID
-
rs6065936
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:46176799 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGATGGCTTCTGGAGCCGAGGCTGG[G/T]AGCTCTGGAGGGGGATGCCCGCGGG
- Phenotype
-
MIM: 609920
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CDH22
PubMed Links
Gene Details
- Gene
- CDH22
- Gene Name
- cadherin 22
View Full Product Details