Product Details
- SNP ID
-
rs6136233
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:17946384 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAATGGCCAGTTTCAAGGCTGGAG[C/T]AGAGAGTAAGATTAGCATGGCCTAT
- Phenotype
-
MIM: 605937
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SNX5
PubMed Links
Gene Details
- Gene
- SNX5
- Gene Name
- sorting nexin 5
View Full Product Details