Product Details

SNP ID
rs2241998
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42576830 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTCTTGTAATTGACAGACTAATG[A/G]GAAGAGCAATGAGATAGAAAGAAAC
Phenotype
MIM: 613429 MIM: 614642
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HAUS2 PubMed Links

Gene Details

Gene
HAUS2
Gene Name
HAUS augmin like complex subunit 2
There are no transcripts associated with this gene.

Gene
STARD9
Gene Name
StAR related lipid transfer domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020759.2 Intron NP_065810.2
XM_011521831.2 Intron XP_011520133.1
XM_011521832.2 Intron XP_011520134.1
XM_011521833.2 Intron XP_011520135.1
XM_011521834.2 Intron XP_011520136.1
XM_011521835.2 Intron XP_011520137.1
XM_011521836.2 Intron XP_011520138.1
XM_011521837.2 Intron XP_011520139.1
XM_011521839.2 Intron XP_011520141.1
XM_017022439.1 Intron XP_016877928.1
XM_017022440.1 Intron XP_016877929.1
XM_017022441.1 Intron XP_016877930.1
XM_017022442.1 Intron XP_016877931.1
XM_017022443.1 Intron XP_016877932.1

View Full Product Details