Product Details

SNP ID
rs10250406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:35636045 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATCCTTGTCAAGTTAGAAAGAA[C/T]TGTGATTCTGAGGTTTGCTTATATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HERPUD2 PubMed Links

Gene Details

Gene
HERPUD2
Gene Name
HERPUD family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022373.4 Intron NP_071768.3
XM_006715765.1 Intron XP_006715828.1
XM_011515473.1 Intron XP_011513775.1
XM_017012506.1 Intron XP_016867995.1

View Full Product Details